Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A1R Antibody, Novus Biologicals™
SDP

Catalog No. NB035544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB035544 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB035544 Supplier Novus Biologicals Supplier No. NBP287922
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Adenosine A1R
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/mL, Flow Cytometry 10 ug/mL
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA
Gene Alias adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1
Host Species Goat
Immunogen Peptide with sequence NDHFRCQPAPPIDED corresponding to C-Terminus according to NP_001041695.1.
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Growth and Development, Immunology, Innate Immunity, Neuronal Cell Markers, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 134
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.