Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2aR/A2bR Antibody (826037), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB295427 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295427 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295427 Supplier Novus Biologicals Supplier No. FAB94973D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenosine A2aR/A2bR Monoclonal antibody specifically detects Adenosine A2aR/A2bR in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Adenosine A2aR/A2bR
Applications Flow Cytometry
Classification Monoclonal
Clone 826037
Conjugate DyLight 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Adenosine A2aR, Met1-Ser412, Accession # P29274
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.