Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2aR Antibody (599717), HRP, Novus Biologicals™
SDP

Catalog No. NB345696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345696 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345696 Supplier Novus Biologicals Supplier No. FAB9497H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenosine A2aR Monoclonal specifically detects Adenosine A2aR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Adenosine A2aR
Applications Flow Cytometry
Classification Monoclonal
Clone 599717
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Adenosine A2a R, Met1-Ser412, Accession # P29274
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.