Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2aR Antibody (7F6-G5-A2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013635 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013635 Supplier Novus Biologicals Supplier No. NBP280562
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenosine A2aR Monoclonal specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen Adenosine A2aR
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7F6-G5-A2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry : 1 - 5 ug/ml, Immunohistochemistry : 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:100. Use reported in scientific literature (PMID 25836920), Immunohistochemistry-Paraffin : 1 - 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 9822147), Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Gene Symbols ADORA2A
Host Species Mouse
Immunogen Full length recombinant human Adenosine A2a R. [UniProt# P29274]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 135
Test Specificity Sheep reactivity reported in scientific literature (PMID: 29791204). Please note that this antibody is reactive to Mouse and derived from the same host, Mouse. Mouse-On-Mouse blocking reagent may be needed for IHC and ICC experiments to reduce high background signal. You can find these reagents under catalog numbers PK-2200-NB and MP-2400-NB. Please contact Technical Support if you have any questions.
Target Species Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.