Learn More
Description
Specifications
Specifications
| Antigen | Adenosine A2aR |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 7F6-G5-A2 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 |
| Gene Symbols | ADORA2A |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
