Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2aR Antibody (7F6-G5-A2), HRP, Novus Biologicals™
SDP

Catalog No. NBP139474H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP139474H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP139474H Supplier Novus Biologicals Supplier No. NBP139474H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenosine A2aR Monoclonal specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Adenosine A2aR
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7F6-G5-A2
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Gene Symbols ADORA2A
Host Species Mouse
Immunogen Full length recombinant human Adenosine A2a R. [UniProt# P29274]
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Human, Mouse, Rat, Pig, Canine, Guinea Pig, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.