Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2aR Antibody, Novus Biologicals™
SDP

Catalog No. NB300597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300597 Supplier Novus Biologicals Supplier No. NB300597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenosine A2aR Polyclonal specifically detects Adenosine A2aR in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Adenosine A2aR
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:20, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:200
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Gene Symbols ADORA2A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(373) S H G D M G L P D V E L L S H E L K(391) of canine A2aAR.
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135
Test Specificity Detects adenosine receptor A2a. This does not detect other AR subtypes.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.