Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2b R Antibody (2683F), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB315136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315136 Supplier Novus Biologicals Supplier No. FAB104442APCCY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Adenosine A2b R Monoclonal specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Adenosine A2b R
Applications Flow Cytometry
Classification Monoclonal
Clone 2683F
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias adenosine A2b receptor, adenosine receptor A2b, ADORA2
Host Species Rabbit
Immunogen Human Adenosine A2b R synthetic peptide, Accession # NP_000667
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 136
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.