Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2b R Antibody (2683F), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340020 Supplier Novus Biologicals Supplier No. FAB104442MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Adenosine A2b R Monoclonal antibody specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Adenosine A2b R
Applications Flow Cytometry
Classification Monoclonal
Clone 2683F
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias adenosine A2b receptor, adenosine receptor A2b, ADORA2
Host Species Rabbit
Immunogen Human Adenosine A2b R synthetic peptide, Accession # NP_000667
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 136
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.