Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2b R Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB202347 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB202347 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB202347 Supplier Novus Biologicals Supplier No. NBP241312G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenosine A2b R Polyclonal antibody specifically detects Adenosine A2b R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Adenosine A2b R
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias adenosine A2b receptor, adenosine receptor A2b, ADORA2
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ADORA2B. The immunogen is located within the last 50 amino acids of ADORA2B. Amino Acid Squence: QADVKSGNGQAGVQPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 136
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.