Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A2b R Antibody, Novus Biologicals™
SDP

Catalog No. NB600797 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600797 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600797 Supplier Novus Biologicals Supplier No. NB600797
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Adenosine A2b R Polyclonal specifically detects Adenosine A2b R in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Adenosine A2b R
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 10ug/ml, Immunohistochemistry 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 2-4ug/ml, Immunohistochemistry-Paraffin 3 ug/ml, Peptide ELISA detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P29275
Gene Alias adenosine A2b receptor, adenosine receptor A2b, ADORA2
Gene Symbols ADORA2B
Host Species Goat
Immunogen Peptide with sequence CQADVKSGNGQ corresponding to C-Terminus according to NP_000667.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 136
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.