Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A3 R Antibody, Novus Biologicals™
SDP

Catalog No. NBP226063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226063 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226063 Supplier Novus Biologicals Supplier No. NBP226063
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Adenosine A3 R Polyclonal specifically detects Adenosine A3 R in Human, Rat, Bovine samples. It is validated for Peptide ELISA.

Specifications

Antigen Adenosine A3 R
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias A3AR, AD026, adenosine A3 receptor, adenosine receptor A3, bA552M11.5
Gene Symbols ADORA3
Host Species Goat
Immunogen Peptide with sequence TASAICNYNAH corresponding to internal region according to NP_065734.5, NP_000668.1, NP_001075445.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 140
Test Specificity This antibody is expected to recognize isoform 1 (NP_065734.5) and isoform 3 (NP_001075445.1).
Target Species Human, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.