Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Deaminase/ADA Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP17771001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17771001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17771001 Supplier Novus Biologicals Supplier No. NBP1777100.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Adenosine Deaminase/ADA
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Accession No. NC_000020.10
Gene Alias ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4
Gene Symbols ADA
Host Species Rabbit
Immunogen Adenosine Deaminase [Calf Spleen]
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100
Reconstitution Restore with 100 uL deionized water (or equivalent)
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.