Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine Deaminase/ADA Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB105657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105657 Supplier Novus Biologicals Supplier No. NBP298771B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenosine Deaminase/ADA Polyclonal specifically detects Adenosine Deaminase/ADA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Adenosine Deaminase/ADA
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Adenosine Deaminase/ADA.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.