Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Rabbit anti-Human, Mouse, Clone: 5R10B4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316575100UL
Description
Adenosine Deaminase/ADA Monoclonal antibody specifically detects Adenosine Deaminase/ADA in Human, Mouse samples. It is validated for Western BlotSpecifications
Adenosine Deaminase/ADA | |
Monoclonal | |
Unconjugated | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Adenosine Deaminase/ADA (P00813). | |
100 μg | |
Cancer, Endocrinology | |
100 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
5R10B4 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction