Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Rabbit anti-Human, Mouse, Clone: 5R10B4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Adenosine Deaminase/ADA |
---|---|
Clone | 5R10B4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Adenosine Deaminase/ADA Monoclonal antibody specifically detects Adenosine Deaminase/ADA in Human, Mouse samples. It is validated for Western BlotSpecifications
Adenosine Deaminase/ADA | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
100 | |
IgG | |
Affinity purified |
5R10B4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Adenosine Deaminase/ADA (P00813). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title