Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus 3 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB362464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362464 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362464 Supplier Novus Biologicals Supplier No. NB10093578F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenovirus 3 Polyclonal antibody specifically detects Adenovirus 3 in Nonspecies specific, Adenovirus, Virus samples. It is validated for Western Blot

Specifications

Antigen Adenovirus 3
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias ad3, adenovirus, Adenovirus 3
Host Species Rabbit
Immunogen Synthetic peptide within residues 250-270 of the fiber knob protein of Ad 3.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific, Adenovirus, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.