Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus-9 E4 Orf1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB344189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344189 Supplier Novus Biologicals Supplier No. NBP241088MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenovirus-9 E4 Orf1 Polyclonal antibody specifically detects Adenovirus-9 E4 Orf1 in Adenovirus, Virus samples. It is validated for ELISA

Specifications

Antigen Adenovirus-9 E4 Orf1
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias 13.6kDa protein, 14.5kDa protein, Control protein orf2, Control protein orf3, Control protein orf4, E4, E4 13.5kDa protein, E4 13.6kDa protein, E4 14.4kDa protein, E4 14.5kDa protein, E4 control protein 34 kDa, E4 control protein orf2, E4 control protein orf3, E4 control protein orf4, HuAdVD_gp17, ORF 2, Orf 3, Orf 4, ORF2, ORF3, ORF3 protein, ORF4, Putative E4
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the Ad-9 E4 Orf1. The immunogen is located within the last 50 amino acids of Adenovirus-9 E4 Orf1. Amino Acid Squence: FNHTDRFLYVREGHPV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6386282
Target Species Adenovirus, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.