Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenovirus Hexon Antibody (1431 (2C4)), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10062300C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062300C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062300C Supplier Novus Biologicals Supplier No. NB10062300C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adenovirus Hexon Monoclonal specifically detects Adenovirus Hexon in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Adenovirus Hexon
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1431 (2C4)
Conjugate DyLight 650
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Hexon protein; Late protein 2; PII
Host Species Mouse
Immunogen ATCC VR5 virus
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62300 recognizes the penton or hexon group antigen present in most adenovirus serotypes.Hexon protein is a major coat protein of adenoviruses. Adenoviruses capsids have three principal protein components: the hexon, the penton, and the fiber. Hexon consists of three subunits together forming two major components of different morphological symmetry. A triangular top with three towers of density is superimposed on a more bulky pseudo hexagonal base.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.