Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Adenovirus Type 2 and 5 Ab-5 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and immunoprecipitation experiments.

$635.00 - $871.00

Specifications

Antigen Adenovirus Type 2 and 5 Ab-5
Clone M58 + M73
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1069P1ABX
View Documents
Epredia™
MS1069P1ABX
Concentrated 100 μL
Each for $635.00
Only null left
Add to Cart
 
MS1069PABX
View Documents
Epredia™
MS1069PABX
Concentrated 200 μL
Each for $871.00
Only null left
Add to Cart
 
MS1069P0
View Documents
Epredia™
MS1069P0
Concentrated 100 μL N/A N/A N/A
MS1069P
View Documents
Epredia™
MS1069P
Concentrated 1 mL N/A N/A N/A
MS1069R1
View Documents
Epredia™
MS1069R1
Prediluted 1 mL N/A N/A N/A
MS1069R7
View Documents
Epredia™
MS1069R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Adenovirus Ab-5 detects adenovirus serogroups 2 and 5. The adenovirus early gene product E1A is a potent stimulator of cellular proliferation, which when overexpressed can overcome the growth-inhibitory effects of TGF-β. The E1A region encodes a series of related proteins (35-46kDa) with multifuctional capabilities and form a specific complex with the retinoblastoma tumor suppressor gene product (Rb protein). The E1A and E1B regions together comprise the transforming region of adenovirus. While expression of E1A alone is sufficient to immortalize primary cells, complete transformation requires the additional expression of the E1B region. Several conserved regions of E1A are similar to portions of other viral oncoproteins like the HPV-16 and HPV-18 E7 and SV40 large T antigen.

Host Species: Mouse

Clone: M58 + M73

Isotype: IgG2a/κ + IgG2a/κ

Species Reactivity: Adenovirus.

Immunogen: Adenovirus

Positive Control: 293 cells or adenovirus infected tissues

Cellular Localization: Nuclear.

Ab-5 is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Adenovirus Type 2 and 5 Ab-5
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Adenovirus
IgG2a κ
M58 + M73
Monoclonal
Mouse
Infectious Disease
Adenovirus
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.