Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase 3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360350 Supplier Novus Biologicals Supplier No. NBP305527APC

Chicken Polyclonal Antibody

Adenylate Cyclase 3 Polyclonal antibody specifically detects Adenylate Cyclase 3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Adenylate Cyclase 3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias AC3EC 4.6.1.1, AC-III, adenylate cyclase 3, adenylate cyclase type 3, Adenylate cyclase type III, Adenylyl cyclase 3, adenylyl cyclase, type III, ATP pyrophosphate-lyase 3, EC 4.6.1, KIAA0511, olfactive type
Host Species Chicken
Immunogen C-terminal peptide of rat Adenlyate Cyclase 3, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 109
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.