Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase 9 Antibody, Novus Biologicals™
SDP

Catalog No. NB437635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB437635 25ul
NBP230380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB437635 Supplier Novus Biologicals Supplier No. NBP23038025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adenylate Cyclase 9 Polyclonal specifically detects Adenylate Cyclase 9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Adenylate Cyclase 9
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O60503
Gene Alias adenylate cyclase type 9, AC9, adenylate cyclase 9
Gene Symbols ADCY9
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: FVMSQVRSRSTFLKVGQSIMHGKDLEVEKALKERMIHSVMPRIIADDLMKQGDEESENSVKRHATSSPKNRKKKSSIQKAPIAFRPFKMQQIEEV
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.