Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Cyclase Toxin Mouse anti-Bacteria, Clone: 9D4, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $456.56
Specifications
Antigen | Adenylate Cyclase Toxin |
---|---|
Clone | 9D4 |
Concentration | Please refer to lot specific datasheet. |
Applications | Dot Blot, Functional Assay, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2087-25
|
MilliporeSigma
MABF208725UL |
25 μL |
Each for $192.35
|
|
|||||
MABF2087-100
|
MilliporeSigma
MABF2087100UL |
100 μL |
Each for $456.56
|
|
|||||
Description
Anti-Adenylate Cyclase Toxin, B. pertussis, clone 9D4 Function Assay and Western Blotting.Specifications
Adenylate Cyclase Toxin | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Bacteria | |
IgG2a λ | |
Protein G Purified | |
Clone 9D4 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis. |
9D4 | |
Dot Blot, Functional Assay, Western Blot | |
Unconjugated | |
Mouse | |
Inflammation and Immunology | |
Adenylate cyclase toxin from Bordetella pertussis. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title