Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Kinase 1 Antibody (7E9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB162335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162335 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162335 Supplier Novus Biologicals Supplier No. NBP259485AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenylate Kinase 1 Monoclonal antibody specifically detects Adenylate Kinase 1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Adenylate Kinase 1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 7000000000
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase
Host Species Mouse
Immunogen Recombinant human Adenylate Kinase 1 (1-194aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.