Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adiponectin/Acrp30 Mouse anti-Mouse, DyLight 594, Clone: MADI 1147, Novus Biologicals™
SDP

Catalog No. NB172160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172160 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172160 Supplier Novus Biologicals Supplier No. NBP280030DL594
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adiponectin/Acrp30 Monoclonal antibody specifically detects Adiponectin/Acrp30 in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen Adiponectin/Acrp30
Applications Western Blot, ELISA
Classification Monoclonal
Clone MADI 1147
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein
Host Species Mouse
Immunogen Recombinant mouse Adiponectin/Acrp30.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.