Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponectin receptor 1 Mouse anti-Human, Clone: 1F12B3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Adiponectin receptor 1 |
---|---|
Clone | 1F12B3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7037
![]() ![]() |
Proteintech Group Inc
666191IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody was raised against 1-72 aa of human AdipoR1.
The adiponectin receptors, ADIPOR1 and ADIPOR2, serve as receptors for globular and full-length adiponectin and mediate increased AMPK and PPAR-alpha ligand activities, as well as fatty acid oxidation and glucose uptake by adiponectin.Specifications
Adiponectin receptor 1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ACDCR1, Adiponectin receptor 1, Adiponectin receptor protein 1, ADIPOR1, CGI 45, CGI45, PAQR1, TESBP1A | |
ADIPOR1 | |
IgG1 | |
Protein G | |
Antibody |
1F12B3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q96A54 | |
51094 | |
Adiponectin receptor 1 Fusion Protein Ag18187 | |
Primary | |
-20°C | |
ADIPOR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title