Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AdipoR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018471 0.025 mg
NBP128641 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018471 Supplier Novus Biologicals Supplier No. NBP1286410.025MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 4 publications

AdipoR2 Polyclonal specifically detects AdipoR2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen AdipoR2
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250
Formulation PBS (pH 7.2) and 1.0 mg/ml BSA with 0.1% Sodium Azide
Gene Accession No. Q86V24
Gene Alias ACDCR2, adiponectin receptor 2, adiponectin receptor protein 2, FLJ21432, PAQR2MGC4640, Progestin and adipoQ receptor family member II
Gene Symbols ADIPOR2
Host Species Goat
Immunogen Synthetic peptide (HFHGVSNLQEFRFMIGGGC) corresponding to aa 362-380 of the C-terminal domain of Human, Mouse and Rat ADPR 2 Receptor
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79602
Target Species Human, Rat, Monkey, Mouse
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.