Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AdipoR2 Goat anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018648 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018648 0.025 mg
NBP147241 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018648 Supplier Novus Biologicals Supplier No. NBP1472410.025MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

AdipoR2 Polyclonal specifically detects AdipoR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AdipoR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250
Formulation 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA, pH 7.2 with 0.1% Sodium Azide
Gene Accession No. Q86V24
Gene Alias ACDCR2, adiponectin receptor 2, adiponectin receptor protein 2, FLJ21432, PAQR2MGC4640, Progestin and adipoQ receptor family member II
Gene Symbols ADIPOR2
Host Species Goat
Immunogen Synthetic peptide (30 amino acid) CSRTPEPDIRLRKGHQLDGTRRGDNDSHQG corresponding to residues 11-40 of the N-terminal domain of human AdipoR2.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79602
Target Species Mouse, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.