Learn More
Bullard™ Adjustable Flow Assemblies for RT Series Airline Respirators
SDP
Flow control assembly for use with RT Series respirator does not include breathing tube.
$126.00 - $137.00
Products 7
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
19-807-856
SDP
Required Accessories View Documents
Bullard™
F40
0.25 inch, industrial interchange
Each for $126.00
Only null left
Add to Cart
19-824-393
SDP
View Documents
Bullard™
F40B
0.25 in., Industrial Interchange Brass
Each for $130.00
Only null left
Add to Cart
19-021-472
SDP
View Documents
Bullard™
F40S
0.25 in., Industrial Interchange Steel
Each for $127.00
Only null left
Add to Cart
19-824-395
SDP
View Documents
Bullard™
F41
0.25 in., Schrader Nipple
Each for $137.00
Only null left
Add to Cart
19-824-396
SDP
View Documents
Bullard™
F42
0.25 in., Snap-Tite Steel Nipple N/A N/A N/A
19-824-397
SDP
View Documents
Bullard™
F44
0.25 in., Snap-Tite Stainless Steel Nipple N/A N/A N/A
19-824-398
SDP
View Documents
Bullard™
F47
0.25 in., CEJN Nipple N/A N/A N/A
Description
Description
  • Adjustable flow accessory
  • For use with RT series airline respirator
  • Flow control assemblies
  • Nipple types include CEJN, industrial interchange, schrader, or snap-tite in brass or steel
  • Does not include breathing tube
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.