Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADK Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB064259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064259 Supplier Novus Biologicals Supplier No. NBP300077V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADK Polyclonal specifically detects ADK in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen ADK
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias adenosine kinase, AKAdenosine 5'-phosphotransferase, EC 2.7.1, EC 2.7.1.20
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ADK (AAH03568.1; Met 1-His 345).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 132
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.