Learn More
Promega ADP-Glo™ Kinase Assay + FES Kinase Enzyme System
Catalog No. PRV9311 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9311 1 Ea.
1 options
Catalog No. PRV9311 Supplier Promega Supplier No. V9311
Only null left
Add to Cart
Add to Cart
Recombinant full-length human FES was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. FES is a proto-oncogene that encodes a protein-tyrosine kinase distinct from c-Src, c-Abl and other nonreceptor tyrosine kinases.
  • Easily Screen and Profile FES Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures
Kit Contents
  • FES Kinase Enzyme System; (V1981) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Product Type Kinase Assay + Kinase Enzyme System