Learn More
Promega ADP-Glo™ Kinase Assay + MET (M1250T) Kinase Enzyme System
Click to view available options
Quantity:
1 Ea.
Description
- Easily Screen and Profile MET (M1250T) Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
Quantity | 1 Ea. |
Content And Storage | Components at different temperatures |
Kit Contents |
|
Product Type | Kinase Assay + Kinase Enzyme System |