Learn More
Description
- Substrate and Assay Reagents for Detecting Class I PI3K Activity
- Use in screening assays with purified PI3K Class I lipid kinases
- Simple, scalable add-and-read assay format
Specifications
Specifications
| Quantity | 1000 Assays |
| Content And Storage | Upon receiving ADP-Glo™ Kinase Assay, remove ATP and store it below -65°C. Store the rest of the components at -30°C to -10°C. Before use, thaw all components completely at room temperature. |
| Format | 384-well Plate |
| Kit Contents |
|
| Product Type | Lipid Kinase Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.