Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ADP-Glo™ Kinase Assay with PI:3PS

Catalog No. p-6837027
Click to view available options
Quantity:
10,000 Assays
1000 Assays

The ADP-Glo™ Kinase assay and lipid kinase substrates PIP2:3PS and PI:3PS are components of the PI3K-Glo™ Class I Kinase Profiling System that are available for purchase separately.

  • Substrate and Assay Reagents for Detecting Class I PI3K Activity
  • Use in screening assays with purified PI3K Class I lipid kinases
  • Simple, scalable add-and-read assay format

Specifications

Quantity 1000 Assays
Content And Storage Upon receiving ADP-Glo™ Kinase Assay, remove ATP and store it below -65°C. Store the rest of the components at -30°C to -10°C. Before use, thaw all components completely at room temperature.
Format 384-well Plate
Kit Contents
  • PI:3PS Substrate, 1mg/mL PI; (V171A) 1 x 0.5mL
  • Lipid Dilution Buffer, 10X; (V180A) 1 x 1mL
  • MgCl2, 1M; (V330A) 1 x 1mL
  • ADP-Glo Reagent; (V912A) 1 x 5mL
  • Kinase Detection Buffer; (V913A) 1 x 10mL
  • Kinase Detection Substrate; (V914A) 1 x 1 each
  • Ultra Pure ATP, 10mM; (V915A) 1 x 500μL
  • ADP, 10mM; (V916A) 1 x 500μL
Product Type Lipid Kinase Assay
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.