Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADP-ribosylarginine hydrolase Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB041722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041722 Supplier Novus Biologicals Supplier No. NBP297042AF405

Rabbit Polyclonal Antibody

ADP-ribosylarginine hydrolase Polyclonal specifically detects ADP-ribosylarginine hydrolase in Human samples. It is validated for Western Blot.

Specifications

Antigen ADP-ribosylarginine hydrolase
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias [Protein ADP-ribosylarginine] hydrolase, ADP-ribosylarginine hydrolaseARH1ADP-ribose-L-arginine cleaving enzyme, EC 3.2.2.19
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ADP-ribosylarginine hydrolase (NP_001116.1; Met1-Leu351).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 141
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.