Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADPRHL2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP188835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP188835 0.1 mL
NB431085 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP188835 Supplier Novus Biologicals Supplier No. NBP188835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADPRHL2 Polyclonal specifically detects ADPRHL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen ADPRHL2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias [Protein ADP-ribosylarginine] hydrolase-like protein 2, ADP-ribosylhydrolase 3, ADP-ribosylhydrolase like 2, ARH3EC 3.2.1.143, FLJ20446, poly(ADP-ribose) glycohydrolase ARH3, protein ADP-ribosylarginine hydrolase-like protein 2
Gene Symbols ADPRHL2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:SEHFLKQLLGHMEDLEGDAQSVLDARELGMEERPYSSRLKKIGELLDQASVTREEVVSELGNGIAAFESVPTAIYCFLR
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 54936
Test Specificity Specificity of human ADPRHL2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.