Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenomedullin/ADM Antibody (OTI8A1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N270100J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270100J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270100J549 Supplier Novus Biologicals Supplier No. NBP270100JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adrenomedullin/ADM Monoclonal specifically detects Adrenomedullin/ADM in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Adrenomedullin/ADM
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI8A1
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias adrenomedullin, AMpreproadrenomedullin
Gene Symbols ADM
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 22-146 of human Adrenomedullin/ADM (NP_001115) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, metabolism, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 133.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.