Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRM1 Antibody (34C2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB160488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160488 Supplier Novus Biologicals Supplier No. NBP259402AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADRM1 Monoclonal antibody specifically detects ADRM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen ADRM1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 34C2
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias 000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog
Host Species Mouse
Immunogen Recombinant human ADRM1 (1-407aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11047
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.