Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADRM1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP188993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP188993 0.1 mL
NB430984 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP188993 Supplier Novus Biologicals Supplier No. NBP188993
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADRM1 Polyclonal specifically detects ADRM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ADRM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q16186
Gene Alias 000 surface antigen, 110 kDa cell membrane glycoprotein, adhesion regulating molecule 1, Adhesion-regulating molecule 1, ARM1, ARM-1, Gp110, GP110hRpn13, M(r) 110, MGC29536, proteasomal ubiquitin receptor ADRM1, Proteasome regulatory particle non-ATPase 13, regulatory particle non-ATPase 13, Rpn13, Rpn13 homolog
Gene Symbols ADRM1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ATMNVPAGPAGGQQVDLASVLTPEIMAPILANADVQERLLPYLPSGESLPQTADEIQNTLTSPQFQQALGMFSAALASGQLGPLMCQFGLPAEAVEAANKGDVEAFAKAMQ
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11047
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.