Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adropin Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP126387G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126387G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126387G Supplier Novus Biologicals Supplier No. NBP126387G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adropin Polyclonal specifically detects Adropin in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Adropin
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q6UWT2
Gene Alias adropin, C9orf165, chromosome 9 open reading frame 165, energy homeostasis associated, Energy homeostasis-associated protein, GAAI470, UNQ470
Gene Symbols ENHO
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Adropin (within residues 10-60). [Swiss-Prot# Q6UWT2]
Molecular Weight of Antigen 8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 375704
Test Specificity Human and mouse. 88% sequence identity with bovine protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.