Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADSL Recombinant Rabbit Monoclonal Antibody (001), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
$310.00 - $593.00
Specifications
| Antigen | ADSL |
|---|---|
| Clone | 001 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543474
|
Invitrogen™
MA543474 |
50 μL |
Each for $310.00
|
|
|||||
PIMA543054
![]() |
Invitrogen™
MA543054 |
100 μL |
Each for $593.00
|
|
|||||
Description
Specificity: Human ADSL/Adenylosuccinate Lyase
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.Specifications
| ADSL | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 2510006M18Rik; Adenylosuccinase; adenylosuccinate lyase; adenylosuccinate lyase 1; Adl; Adsl; AMPS; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; ASase; ASL; null; test; unnamed protein product | |
| Adsl | |
| IgG | |
| Protein A | |
| Antibody |
| 001 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P30566 | |
| 158 | |
| Recombinant Human ADSL/Adenylosuccinate Lyase protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Adsl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title