Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADTB1 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB057826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057826 Supplier Novus Biologicals Supplier No. NBP299079IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADTB1 Polyclonal specifically detects ADTB1 in Human samples. It is validated for Western Blot.

Specifications

Antigen ADTB1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Adapter-Related Protein Complex 1 Subunit Beta-1, Adaptor Protein Complex AP-1 Subunit Beta-1, Adaptor-Related Protein Complex 1, Beta 1 Subunit, AP-1 Complex Subunit Beta-1, AP105A, AP1B1, BAM22, beta1-adaptin, Beta-1-adaptin, Beta-Adaptin 1, beta-prime-adaptin, CLAPB2, Clathrin Assembly Protein Complex 1 Beta Large Chain, Golgi Adaptor HA1/AP1 Adaptin Beta Subunit, Plasma Membrane Adaptor HA2/AP2 Adaptor Beta Subunit
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ADTB1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers
Primary or Secondary Primary
Gene ID (Entrez) 162
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.