Learn More
MSA™ Advantage™ 900 Elastomeric Half-Mask Respirators
An elastomeric half-mask respirator providing healthcare professionals, essential workers and first responders a secure protection solution with enhanced communication.
$82.97 - $84.02
Specifications
Connection Type Bayonet
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
With Exhalation Valve No
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-003-388
View Documents
MSA™
10218527
Small
Each for $84.02
Only null left
Add to Cart
19-003-389
View Documents
MSA™
10218528
Medium
Each for $84.02
Only null left
Add to Cart
19-003-390
View Documents
MSA™
10218529
Large
Each for $82.97
Only null left
Add to Cart
Description
Description
The Advantage 900 Elastomeric half-mask respirator provides healthcare professionals, essential workers and first responders a secure protection solution with enhanced communication.
Elastomeric respirators are designed for repeated cleaning and decontamination, resulting in dramatic cost savings over filtering facepiece respirators. The included mechanical speaking diaphragm allows for improved communication among essential workers, patients and the public.
  • No exhalation valve, allowing for filtration of exhaled air and providing a means of source control and use in a sterile field environment
  • Reuse and cleaning capability make for a more eco-friendly alternative
  • Overmolded MultiFlex nosecup provides stability on user's face while maintaining a secure fit
  • Anthrocurve facepiece design maximizes fit and comfort and reduces potential leak points
Specifications
Specifications
Bayonet
Thermoplastic Elastomer
No
Non-sterile
Blue
Half-Mask Respirator
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.