Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ADX 10059 hydrochloride
GREENER_CHOICE

Catalog No. 441610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-161-0 10 mg
44-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-161-0 Supplier Tocris Bioscience™ Supplier No. 4416/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Negative allosteric modulator at mGlu5

ADX 10059 hydrochloride is reported to be a negative allosteric modulator at the mGlu5 receptor. Analog of MPEP (Cat. No. 1212).

Chemical Identifiers

CAS 757949-98-7
Molecular Formula C15H14ClFN2

Specifications

Quantity 10 mg
Formula Weight 276.74
Purity >99%
Molecular Weight (g/mol) 276.739
Chemical Name or Material ADX 10059 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.