Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AE Binding Protein 1/ACLP Antibody (590831) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. NB376594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376594 Supplier Novus Biologicals Supplier No. FAB6425J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AE Binding Protein 1/ACLP Monoclonal antibody specifically detects AE Binding Protein 1/ACLP in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen AE Binding Protein 1/ACLP
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 590831
Conjugate Janelia Fluor 646
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ACLP, ACLPadipocyte enhancer binding protein 1, AE binding protein 1, AE-binding protein 1adipocyte enhancer-binding protein 1, Aortic carboxypeptidase-like protein, FLJ33612
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ACLP, Glu345-Phe1158, Accession # Q8IUX7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.