Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AEBP2 Antibody (2D7B6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61822 Supplier Novus Biologicals Supplier No. NBP261822
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AEBP2 Monoclonal specifically detects AEBP2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen AEBP2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2D7B6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation PBS with 0.05% Sodium Azide
Gene Alias Adipocyte enhancer-binding protein 2, AE binding protein 2, AE(adipocyte enhancer)-binding protein 2, AE-binding protein 2, MGC17922, zinc finger protein AEBP2
Gene Symbols AEBP2
Host Species Mouse
Immunogen Purified recombinant fragment of human AEBP2 (AA: 358-495) expressed in E. Coli.
Molecular Weight of Antigen 54.5 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 121536
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.