Learn More
Description
Specifications
Specifications
| Antigen | AF4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS & 2% Sucrose. with No Preservative |
| Gene Alias | AF-4, AF4/FMR2 family member 1, AF4/FMR2 family, member 1, AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 2, ALL1-fused gene from chromosome 4 protein, FEL, MLLT2MGC134969, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 2, PBM1, pre-B-cell monocytic leukemia partner 1, Protein AF-4, Protein FEL, Proto-oncogene AF4 |
| Gene Symbols | Aff1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to Aff1 (AF4/FMR2 family, member 1) The peptide sequence was selected from the N terminal of Aff1. Peptide sequence MAAHSSLYNEDRNLLRIREKERRNQEAHQEKEAFPEKAPLFPEPYKTAKG. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
