Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Afadin/AF-6 Antibody (3), Novus Biologicals™
SDP

Catalog No. NB119812 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119812 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119812 Supplier Novus Biologicals Supplier No. NBP311187
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Afadin/AF-6 Monoclonal specifically detects Afadin/AF-6 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Afadin/AF-6
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone 3
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias AF64, afadin, FLJ34371, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4
Host Species Mouse
Immunogen Monoclonal antibody 3 has been raised against peptide 1814-1829 aa of rat l-afadin.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4301
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.