Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP Antibody (16F50), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP222004X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222004X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222004X Supplier Novus Biologicals Supplier No. NBP222004AF488

Mouse Monoclonal Antibody

AFAP Monoclonal specifically detects AFAP in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen AFAP
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 16F50
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, ELISA
Gene Accession No. Q12884
Gene Alias actin filament associated protein 1, FLJ56849
Gene Symbols AFAP1
Host Species Mouse
Immunogen Human recombinant AFAP
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60312
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.