Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB165309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165309 Supplier Novus Biologicals Supplier No. NBP281782AF405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AFAP Polyclonal antibody specifically detects AFAP in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen AFAP
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias actin filament associated protein 1, FLJ56849
Host Species Rabbit
Immunogen AFAP antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human AFAP. The immunogen is located within amino acids 30 - 80 of AFAP. Amino Acid Squence: FDVKDHAQKQETANSLPA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 60312
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.